missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Fucose-1-phosphate guanylyltransferase Protein

A DNA sequence encoding the Homo sapiens (Human) Fucose-1-phosphate guanylyltransferase, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP9143-ye-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16057701

  • 277.00€ / 50 microgramos


Fecha estimada de envío
19-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_001186257.2
Fucose-1-phosphate guanylyltransferase Protein
50 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
8790
Greater than 90% as determined by SDS-PAGE.
Research Use Only
FPGT
Recombinant Protein
Yeast
MAAARDPPEV SLREATQRKL RRFSELRGKL VARGEFWDIV AITAADEKQE LAYNQQLSEK LKRKELPLGV QYHVFVDPAG AKIGNGGSTL CALQCLEKLY GDKWNSFTIL LIHSGGYSQR LPNASALGKI FTALPLGNPI YQMLELKLAM YIDFPLNMNP GILVTCADDI ELYSIGEFEF IRFDKPGFTA LAHPSSLTIG TTHGVFVLDP FDDLKHRDLE YRSCHRFLHK PSIEKMYQFN AVCRPGNFCQ QDFAGGDIAD LKLDSDYVYT DSLFYMDHKS AKMLLAFYEK IGTLSCEIDA YGDFLQALGP GATVEYTRNT SNVIKEESEL VEMRQRIFHL LKGTSLNVVV LNNSKFYHIG TTEEYLFYFT SDNSLKSELG LQSITFSIFP DIPECSGKTS CIIQSILDSR CSVAPGSVVE YSRLGPDVSV GENCIISGSY ILTKAALPAH SFVCSLSLKM NRCLKYATMA FGVQDNLKKS VKTLSDIKLL QFFGVCFLSC LDVWNLKVTE ELFSGNKTCL SLWTARIFPV CSSLSDSVIT SLKMLNAVKN KSAFSLNSYK LLSIEEMLIY KDVEDMITYR EQIFLEISLK SSLM
Videos
Documentos