missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FSH Protein
A cDNA sequence encoding the FSH was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10620-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
FSH Protein | |
Research Use Only | |
The ED50 as determined by cAMP accumulation in human FSH Receptor transfected Chinese Hamster Ovary cells was found to be 80-450 pg/ml. | |
Human | |
Untagged | |
The recombinant FSH was lyophilized from a concentrated (1 mg/ml) solution containing PBS. |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
Human (HEK293) | |
FSH subunit alpha: APDVQDCPECTLQENPFFSQPGAPILQCMGCCFSRAYPTPLRSKKTMLVQKNVTSESTCCVAKSYNRVTVMGGFKVENHTACHCSTCYYHKSFSH subunit beta: NSCELTNITIAIEKEECRFCISINTTWCAGYCYTRDLVYKDPARPKIQKTCTFKELVYETVRVPGCAHHADSLYTYPVATQCHCGKCDSDSTDCTVRGLGPSYCSFGEMKE | |
Greater than 95% as determined by SDS-PAGE. |