missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Fructose-1,6-bisphosphatase 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Fructose-1,6-bisphosphatase 1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6023-ec-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15929429

  • 1180.00€ / 500 microgramos


Fecha estimada de envío
06-08-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_000498.2
Fructose-1,6-bisphosphatase 1 Protein
500 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
2203
Greater than 80% as determined by SDS-PAGE.
Research Use Only
FBP1
Recombinant Protein
E. coli
ADQAPFDTDV NTLTRFVMEE GRKARGTGEL TQLLNSLCTA VKAISSAVRK AGIAHLYGIA GSTNVTGDQV KKLDVLSNDL VMNMLKSSFA TCVLVSEEDK HAIIVEPEKR GKYVVCFDPL DGSSNIDCLV SVGTIFGIYR KKSTDEPSEK DALQPGRNLV AAGYALYGSA TMLVLAMDCG VNCFMLDPAI GEFILVDKDV KIKKKGKIYS LNEGYARDFD PAVTEYIQRK KFPPDNSAPY GARYVGSMVA DVHRTLVYGG IFLYPANKKS PNGKLRLLYE CNPMAYVMEK AGGMATTGKE AVLDVIPTDI HQRAPVILGS PDDVLEFLKV YEKHSAQ
Videos
Documentos