missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Fractalkine / CX3CL1 Protein

A DNA sequence encoding the Homo sapiens (Human) Fractalkine / CX3CL1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5896-ec-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15918519

  • 1926.00€ / 1 miligramo


Fecha de envío estimada
del proveedor 05-04-2019
Añadir a la cesta

Especificación

Especificación

NP_001291321.1
Fractalkine / CX3CL1 Protein
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
6376
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CX3CL1
Recombinant Protein
E. coli
QHHGVTKCNI TCSKMTSKIP VALLIHYQQN QASCGKRAII LETRQHRLFC ADPKEQWVKD AMQHLDRQAA ALTRNG
Videos