missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Fos-related antigen 2 Protein

A DNA sequence encoding the Homo sapiens (Human) Fos-related antigen 2, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6053-ec-100ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15909649

  • 437.00€ / 100 microgramos


Fecha estimada de envío
12-08-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_005244.1
Fos-related antigen 2 Protein
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
2355
Greater than 90% as determined by SDS-PAGE.
Research Use Only
FOSL2
Recombinant Protein
E. coli
MYQDYPGNFD TSSRGSSGSP AHAESYSSGG GGQQKFRVDM PGSGSAFIPT INAITTSQDL QWMVQPTVIT SMSNPYPRSH PYSPLPGLAS VPGHMALPRP GVIKTIGTTV GRRRRDEQLS PEEEEKRRIR RERNKLAAAK CRNRRRELTE KLQAETEELE EEKSGLQKEI AELQKEKEKL EFMLVAHGPV CKISPEERRS PPAPGLQPMR SGGGSVGAVV VKQEPLEEDS PSSSSAGLDK AQRSVIKPIS IAGGFYGEEP LHTPIVVTST PAVTPGTSNL VFTYPSVLEQ ESPASPSESC SKAHRRSSSS GDQSSDSLNS PTLLAL
Videos