missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Forkhead box protein M1 Protein

A DNA sequence encoding the Homo sapiens (Human) Forkhead box protein M1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6054-ec-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15919659

  • 310.00€ / 50 microgramos


Fecha estimada de envío
16-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_001230017.1
Forkhead box protein M1 Protein
50 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
2305
Greater than 80% as determined by SDS-PAGE.
Research Use Only
FOXM1
Recombinant Protein
E. coli
ERPPYSYMAM IQFAINSTER KRMTLKDIYT WIEDHFPYFK HIAKPGWKNS IRHNLSLHDM FVRETSANGK VSFWTIHPSA NRYLTLDQVF KPL
Videos