missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Forkhead box protein M1 Protein
A DNA sequence encoding the Homo sapiens (Human) Forkhead box protein M1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6054-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001230017.1 | |
Forkhead box protein M1 Protein | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
2305 | |
Greater than 80% as determined by SDS-PAGE. | |
Research Use Only | |
FOXM1 | |
Recombinant Protein | |
E. coli | |
ERPPYSYMAM IQFAINSTER KRMTLKDIYT WIEDHFPYFK HIAKPGWKNS IRHNLSLHDM FVRETSANGK VSFWTIHPSA NRYLTLDQVF KPL |