missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FN3KRP Protein
A cDNA sequence encoding the FN3KRP was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11894-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
FN3KRP Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMEELLRR ELGCSSVRAT GHSGGGCISQ GRSYDTDQGR VFVKVNPKAE ARRMFEGEMA SLTAILKTNT VKVPKPIKVL DAPGGGSVLV MEHMDMRHLS SHAAKLGAQL ADLHLDNKKL GEMRLKEAGT VGRGGGQEER PFVARFGFDV VTCCGYLPQVNDWQEDWVVF YARQRIQPQM DMVEKESGDR EALQLWSALQ LKIPDLFRDL EIIPALLHGD LWGGNVAEDS SGPVIFDPAS FYGHSEYELA IAGMFGGFSS SFYSAYHGKI PKAPGFEKRL QLYQLFHYLN HWNHFGSGYR GSSLNIMRNL VK | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
FN3KRP protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M urea and 10% glycerol. |