missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human FN3K Protein

A cDNA sequence encoding the FN3K was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP11893-2ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15918718

  • 103.00€ / 2 microgramos


Fecha de envío estimada
del proveedor 10-04-2019
Añadir a la cesta

Especificación

Especificación

FN3K Protein
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
FN3K protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT.
Greater than 85.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMEQLLRA ELRTATLRAF GGPGAGCISE GRAYDTDAGP VFVKVNRRTQ ARQMFEGEVA SLEALRSTGL VRVPRPMKVI DLPGGGAAFV MEHLKMKSLS SQASKLGEQM ADLHLYNQKL REKLKEEENT VGRRGEGAEP QYVDKFGFHT VTCCGFIPQV NEWQDDWPTF FARHRLQAQL DLIEKDYADR EARELWSRLQ VKIPDLFCGL EIVPALLHGD LWSGNVAEDD VGPIIYDPAS FYGHSEFELA IALMFGGFPR SFFTAYHRKI PKAPGFDQRL LLYQLFNYLN HWNHFGREYR SPSLGTMRRL LK
Videos