missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human FKBP5 Protein

A cDNA sequence encoding the FKBP5 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP11886-2ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15908698

  • 107.00€ / 2 microgramos


Fecha estimada de envío
25-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

2289
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
FKBP5
Human
His
FKBP5 was filtered (0.4microm) and lyophilized in 20mM Tris buffer and 50mM NaCl, pH 7.5.
FKBP5 Protein
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MKHHHHHHASMTTDEGAKNN EESPTATVAE QGEDITSKKD RGVLKIVKRV GNGEETPMIG DKVYVHYKGK LSNGKKFDSS HDRNEPFVFS LGKGQVIKAW DIGVATMKKG EICHLLCKPE YAYGSAGSLP KIPSNATLFF EIELLDFKGE DLFEDGGIIR RTKRKGEGYS NPNEGATVEI HLEGRCGGRM FDCRDVAFTV GEGEDHDIPI GIDKALEKMQ REEQCILYLG PRYGFGEAGK PKFGIEPNAE LIYEVTLKSF EKAKESWEMD TKEKLEQAAI VKEKGTVYFK GGKYMQAVIQ YGKIVSWLEM EYGLSEKESK ASESFLLAAF LNLAMCYLKL REYTKAVECC DKALGLDSAN EKGLYRRGEA QLLMNEFESA KGDFEKVLEV NPQNKAARLQ ISMCQKKAKE HNERDRRIYA NMFKKFAEQD AKEEANKAMG KKTSEGVTNE KGTDSQAMEE EKPEGHV
Videos
Documentos