missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Fin bud initiation factor homolog Protein
A DNA sequence encoding the Homo sapiens (Human) Fin bud initiation factor homolog, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7973-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_976249.1 | |
Fin bud initiation factor homolog Protein | |
Research Use Only | |
FIBIN | |
Recombinant Protein | |
E. coli | |
YFDGPLYPEM SNGTLHHYFV PDGDYEENDD PEKCQLLFRV SDHRRCSQGE GSQVGSLLSL TLREEFTVLG RQVEDAGRVL EGISKSISYD LDGEESYGKY LRRESHQIGD AYSNSDKSLT ELESKFKQGQ EQDSRQESRL NEDFLGMLVH TRSLLKETLD ISVGLRDKYE LLALTIRSHG TRLGRLKNDY LKV | |
Greater than 90% as determined by SDS-PAGE. |
387758 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |