missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Fibroblast growth factor 19 Protein

A DNA sequence encoding the Human Fibroblast growth factor 19, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10383-ec-5ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15923688

Cantidad Precio
1 105.00€ / 5 microgramos
10 98.00€ / 5 microgramos
Fecha estimada de envío
07-09-2020
Añadir a la cesta


Especificaciones

Especificaciones

NP_005108.1
Fibroblast growth factor 19 Protein
5 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
MRPLAFSDAG PHVHYGWGDP IRLRHLYTSG PHGLSSCFLR IRADGVVDCA RGQSAHSLLE IKAVALRTVA IKGVHSVRYL CMGADGKMQG LLQYSEEDCA FEEEIRPDGY NVYRSEKHRL PVSLSSAKQR QLYKNRGFLP LSHFLPMLPM VPEEPEDLRG HLESDMFSSP LETDSMDPFG LVTGLEAVRS PSFEK
9965
Greater than 95% as determined by SDS-PAGE and HPLC.
Research Use Only
FGF19
The ED50 as determined by a cell proliferation assay using murine Balb/c 3T3 cells is less than 150 ng/ml, corresponding to a specific activity of > 6.7 M 103 IU/mg.
Human
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos
Documentos