missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human FGFR4 Protein

A cDNA sequence encoding the FGFR4 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP10606-2ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15914398

  • 137.00€ / 2 microgramos
Fecha estimada de envío: 29-05-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Especificaciones

Especificaciones

2264
2 μg
< 1.0 EU per ug protein as determined by the LAL method.
Determined by its ability to inhibit human FGF acidic-dependent proliferation on R1 cells. The ED50 for this effect is typically at 15.0-30.0 ng/ml.
Human
Untagged
CD334 was lyophilized from a concentrated (1 mg/ml) sterile solution containing no additives.
FGFR4 Protein
Research Use Only
FGFR4
Recombinant Protein
Baculovirus
MRLLLALLGV LLSVPGPPVL SLEASEEVEL EPCLAPSLEQ QEQELTVALG QPVRLCCGRA ERGGHWYKEG SRLAPAGRVR GWRGRLEIAS FLPEDAGRYL CLARGSMIVL QNLTLITGDS LTSSNDDEDP KSHRDPSNRH SYPQQAPYWT HPQRMEKKLH AVPAGNTVKF RCPAAGNPTP TIRWLKDGQA FHGENRIGGI RLRHQHWSLV MESVVPSDRG TYTCLVENAV GSIRYNYLLD VLERSPHRPI LQAGLPANTT AVVGSDVELL CKVYSDAQPH IQWLKHIVIN GSSFGADGFP YVQVLKTADI NSSEVEVLYL RNVSAEDAGE YTCLAGNSIG LSYQSAWLTV LPEEDPTWTA AAPEARYTDI ILYASGSLAL AVLLLLAGLY RGQALHGRHP RPPATVQKLS RFPLARQFSL ESGSSGKSSS SLVRGVRLSS SGPALLAGLV SLDLPLDPLW EFPRDRLVLG KPLGEGCFGQ VVRAEAFGMD PARPDQASTV AVKMLKDNAS DKDLADLVSE MEVMKLIGRH KNIINLLGVC TQEGPLYVIV ECAAKGNLRE FLRARRPPGP DLSPDGPRSS EGPLSFPVLV SCAYQVARGM QYLESRKCIH RDLAARNVLV TEDNVMKIAD FGLARGVHHI DYYKKTSNGR LPVKWMAPEA LFDRVYTHQS DVWSFGILLW EIFTLGGSPY PGIPVEELFS LLREGHRMDR PPHCPPELYG LMRECWHAAP SQRPTFKQLV EALDKVLLAV SEEYLDLRLT FGPYSPSGGD ASSTCSSSDS VFSHDPLPLG SSSFPFGSGV QT
Greater than 90.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales