missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FGFR3 / CD333 Protein
A DNA sequence encoding the Homo sapiens (Human) FGFR3 / CD333, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6047-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000133.1 | |
FGFR3 / CD333 Protein | |
Research Use Only | |
FGFR3 | |
Recombinant Protein | |
E. coli | |
RLRSPPKKGL GSPTVHKISR FPLKRQVSLE SNASMSSNTP LVRIARLSSG EGPTLANVSE LELPADPKWE LSRARLTLGK PLGEGCFGQV VMAEAIGIDK DRAAKPVTVA VKMLKDDATD KDLSDLVSEM EMMKMIGKHK NIINLLGACT QGGPLYVLVE YAAKGNLREF LRARRPPGLD YSFDTCKPPE EQLTFKDLVS CAYQVARGME YLASQKCIHR DLAARNVLVT EDNVMKIADF GLARDVHNLD YYKKTTNGRL PVKWMAPEAL FDRVYTHQSD VWSFGVLLWE IFTLGGSPYP GIPVEELFKL LKEGHRMDKP ANCTHDLYMI MRECWHAAPS QRPTFKQLVE DLDRVLTVTS TDEYLDLSAP FEQYSPGGQD TPSSSSSGDD SVFAHDLLPP APPSSGGSRT | |
Greater than 80% as determined by SDS-PAGE. |
2261 | |
200 μg | |
Not determined. | |
Cytoplasmic Domain | |
Human | |
His-B2M | |
Tris-based buffer, 50% glycerol |