missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human FGFR3 / CD333 Protein

A DNA sequence encoding the Homo sapiens (Human) FGFR3 / CD333, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6047-ye-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15909609

  • 310.00€ / 50 microgramos


Fecha estimada de envío
22-11-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_000133.1
FGFR3 / CD333 Protein
50 μg
Not determined.
Cytoplasmic Domain
Human
His-B2M
Tris-based buffer, 50% glycerol
2261
Greater than 80% as determined by SDS-PAGE.
Research Use Only
FGFR3
Recombinant Protein
Yeast
RLRSPPKKGL GSPTVHKISR FPLKRQVSLE SNASMSSNTP LVRIARLSSG EGPTLANVSE LELPADPKWE LSRARLTLGK PLGEGCFGQV VMAEAIGIDK DRAAKPVTVA VKMLKDDATD KDLSDLVSEM EMMKMIGKHK NIINLLGACT QGGPLYVLVE YAAKGNLREF LRARRPPGLD YSFDTCKPPE EQLTFKDLVS CAYQVARGME YLASQKCIHR DLAARNVLVT EDNVMKIADF GLARDVHNLD YYKKTTNGRL PVKWMAPEAL FDRVYTHQSD VWSFGVLLWE IFTLGGSPYP GIPVEELFKL LKEGHRMDKP ANCTHDLYMI MRECWHAAPS QRPTFKQLVE DLDRVLTVTS TDEYLDLSAP FEQYSPGGQD TPSSSSSGDD SVFAHDLLPP APPSSGGSRT
Videos
Documentos