missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human FGFBP Protein

A cDNA sequence encoding the FGFBP was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP11873-2ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15918658

  • 103.00€ / 2 microgramos


Fecha estimada de envío
22-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

FGFBP Protein
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
FGFBP protein solution (0.25 mg/ml) containing Phosphate buffered saline (pH 7.4), 10% glycerol and 1mM DTT.
Greater than 85.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSKKKVKNG LHSKVVSEQK DTLGNTQIKQ KSRPGNKGKF VTKDQANCRW AATEQEEGIS LKVECTQLDH EFSCVFAGNP TSCLKLKDER VYWKQVARNL RSQKDICRYS KTAVKTRVCR KDFPESSLKL VSSTLFGNTK PRKEKTEMSP REHIKGKETT PSSLAVTQTM ATKAPECVED PDMANQRKTA LEFCGETWSS LCTFFLSIVQ DTSC
Videos