missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FGF7 / FGF-7 / KGF Protein
A DNA sequence encoding the Human FGF7 / FGF-7 / KGF, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10379-ec-250ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_002000.1 | |
FGF7 / FGF-7 / KGF Protein | |
Research Use Only | |
FGF7 | |
The ED50 as determined by thymidine uptake assay using FGF-receptors transfected BaF3 cells is less than 10 ng/ml, corresponding to a specific activity of > 1.0 M 105 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered 20 mM PBS, 0.5 M NaCl, pH 8.0, lyophilized |
2252 | |
250 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
CNDMTPEQMA TNVNCSSPER HTRSYDYMEG GDIRVRRLFC RTQWYLRIDK RGKVKGTQEM KNNYNIMEIR TVAVGIVAIK GVESEFYLAM NKEGKLYAKK ECNEDCNFKE LILENHYNTY ASAKWTHNGG EMFVALNQKG IPVRGKKTKK EQKTAHFLPM AIT | |
Greater than 96% as determined by SDS-PAGE and HPLC. |