missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human FGF7 / FGF-7 / KGF Protein

A DNA sequence encoding the Human FGF7 / FGF-7 / KGF, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10379-ec-100ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15913678

  • 1141.00€ / 100 microgramos


Fecha de envío estimada
del proveedor 21-05-2019
Añadir a la cesta

Especificación

Especificación

NP_002000.1
FGF7 / FGF-7 / KGF Protein
100 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
CNDMTPEQMA TNVNCSSPER HTRSYDYMEG GDIRVRRLFC RTQWYLRIDK RGKVKGTQEM KNNYNIMEIR TVAVGIVAIK GVESEFYLAM NKEGKLYAKK ECNEDCNFKE LILENHYNTY ASAKWTHNGG EMFVALNQKG IPVRGKKTKK EQKTAHFLPM AIT
2252
Greater than 96% as determined by SDS-PAGE and HPLC.
Research Use Only
FGF7
The ED50 as determined by thymidine uptake assay using FGF-receptors transfected BaF3 cells is less than 10 ng/ml, corresponding to a specific activity of > 1.0 M 105 IU/mg.
Human
Untagged
0.2 um filtered 20 mM PBS, 0.5 M NaCl, pH 8.0, lyophilized
Videos