Learn More
enQuireBio™ Recombinant Human FGF10 / KGF2 Protein
Recombinant Protein
Marca:  enQuireBio™ QP5311-500ug
Detalles adicionales : Peso : 0.01000kg
 Ver más versiones de este producto
Código de producto. 15935899
  • 2025.00€ / 500 microgramos
Fecha estimada de envío
26-03-2020
Add to basket
Specifications
Specifications
NP_004456.1
FGF10 / KGF2 Protein
500 μg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
4MBr-5 cell proliferation is significantly induced by less than 200 ng/ml of human FGF10 or activity measured must be greater than 5.0 x 10^3 units/mg.
Human
Untagged
10 mM sodium phosphate, pH 7.5
2255
Greater than 95% as determined by SDS-PAGE.
Research Use Only
FGF10
Recombinant Protein
E. coli
MLGQDMVSPE . ATNSSSSSF S. SPSSAGRH VR. SYNHLQG DVR. WRKLFS FTKY. FLKIE KNGKV. SGTK KENCPY. SIL EITSVEIGVV AVKAINS. NY YLAMNKKG. K LYGSKEFNN.  DCKLKERIEE . NGYNTYASF N. WQHNGRQM YV. ALNGKGA PRR. GQKTRR KNTS. AHFLP MVVHS
Videos
Documents