missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Ferritin heavy chain Protein

A DNA sequence encoding the Homo sapiens (Human) Ferritin heavy chain, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8397-ec-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16054401

  • 234.00€ / 50 microgramos


Fecha estimada de envío
12-12-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_002023.2
Ferritin heavy chain Protein
50 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
2495
Greater than 90% as determined by SDS-PAGE.
Research Use Only
FTH1
Recombinant Protein
E. coli
TTASTSQVRQ NYHQDSEAAI NRQINLELYA SYVYLSMSYY FDRDDVALKN FAKYFLHQSH EEREHAEKLM KLQNQRGGRI FLQDIKKPDC DDWESGLNAM ECALHLEKNV NQSLLELHKL ATDKNDPHLC DFIETHYLNE QVKAIKELGD HVTNLRKMGA PESGLAEYLF DKHTLGDSDN ES
Videos
Documentos