missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FDXR, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) FDXR, mitochondrial, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6033-ec-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001244941.2 | |
FDXR, mitochondrial Protein | |
Research Use Only | |
FDXR | |
Recombinant Protein | |
E. coli | |
STQEKTPQIC VVGSGPAGFY TAQHLLKQHP QAHVDIYEKQ PVPFGLVRFG VAPDHPEVKS YGAEDHRALE IPGEELPGVC SARAFVGWYN GLPENQELEP DLSCDTAVIL GQGNVALDVA RILLTPPEHL ERTDITKAAL GVLRQSRVKT VWLVGRRGPL QVAFTIKELR EMIQLPGARP ILDPVDFLGL QDKIKEVPRP RKRLTELLLR TATEKPGPAE AARQASASRA WGLRFFRSPQ QVLPSPDGRR AAGVRLAVTR LEGVDEATRA VPTGDMEDLP CGLVLSSIGY KSRPVDPSVP FDSKLGVIPN VEGRVMDVPG LYCSGWVKRG PTGVIATTMT DSFLTGQMLL QDLKAGLLPS GPRPGYAAIQ ALLSSRGVRP VSFSDWEKLD AEEVARGQGT GKPREKLVDP QEMLRLLGH | |
Greater than 80% as determined by SDS-PAGE. |
2232 | |
1 mg | |
Not determined. | |
Full Length of Isoform 6 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |