missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human FCGR3B Protein

A cDNA sequence encoding the FCGR3B was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP11861-2ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15928618

  • 107.00€ / 2 microgramos


Fecha estimada de envío
26-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

2215
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
FCGR3B
Human
His
FCGR3B protein solution (0.25 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol.
FCGR3B Protein
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
Insect
EDLPKAVVFL EPQWYSVLEK DSVTLKCQGA YSPEDNSTQW FHNENLISSQ ASSYFIDAAT VNDSGEYRCQ TNLSTLSDPV QLEVHIGWLL LQAPRWVFKE EDPIHLRCHS WKNTALHKVT YLQNGKDRKY FHHNSDFHIP KATLKDSGSY FCRGLVGSKN VSSETVNITI TQGLAVSTIS VEHHHHHH
Videos
Documentos