missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FCGR3B Protein
A cDNA sequence encoding the FCGR3B was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11861-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
2215 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
EDLPKAVVFL EPQWYSVLEK DSVTLKCQGA YSPEDNSTQW FHNENLISSQ ASSYFIDAAT VNDSGEYRCQ TNLSTLSDPV QLEVHIGWLL LQAPRWVFKE EDPIHLRCHS WKNTALHKVT YLQNGKDRKY FHHNSDFHIP KATLKDSGSY FCRGLVGSKN VSSETVNITI TQGLAVSTIS VEHHHHHH | |
Greater than 90.0% as determined by SDS-PAGE. |
FCGR3B Protein | |
Research Use Only | |
FCGR3B | |
Human | |
His | |
FCGR3B protein solution (0.25 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |