missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FCGR2B Protein
A cDNA sequence encoding the FCGR2B was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11859-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
2213 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMAPPKA VLKLEPQWIN VLQEDSVTLT CRGTHSPESD SIQWFHNGNL IPTHTQPSYR FKANNNDSGE YTCQTGQTSL SDPVHLTVLS EWLVLQTPHL EFQEGETIVL RCHSWKDKPL VKVTFFQNGK SKKFSRSDPN FSIPQANHSH SGDYHCTGNI GYTLYSSKPV TITVQAP | |
Greater than 95.0% as determined by SDS-PAGE. |
FCGR2B Protein | |
Research Use Only | |
FCGR2B | |
Human | |
His | |
FCGR2B protein solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 30% glycerol and 1mM DTT. |