missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FCER1G Protein
A DNA sequence encoding the Homo sapiens (Human) FCER1G, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6027-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_004097.1 | |
FCER1G Protein | |
Research Use Only | |
FCER1G | |
Recombinant Protein | |
E. coli | |
LGEPQLCYIL DAILFLYGIV LTLLYCRLKI QVRKAAITSY EKSDGVYTGL STRNQETYET LKHEKPPQ | |
Greater than 90% as determined by SDS-PAGE. |
2207 | |
200 μg | |
Not determined. | |
Full Length of Mature Protein | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |