missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FBP2 Protein
A cDNA sequence encoding the FBP2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11855-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
FBP2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMTDRSPF ETDMLTLTRY VMEKGRQAKG TGELTQLLNS MLTAIKAISS AVRKAGLAHL YGIAGSVNVT GDEVKKLDVL SNSLVINMVQ SSYSTCVLVS EENKDAIITA KEKRGKYVVC FDPLDGSSNI DCLASIGTIF AIYRKTSEDE PSEKDALQCG RNIVAAGYAL YGSATLVALS TGQGVDLFML DPALGEFVLV EKDVKIKKKG KIYSLNEGYA KYFDAATTEY VQKKKFPEDG SAPYGARYVG SMVADVHRTL VYGGIFLYPA NQKSPKGKLR LLYECNPVAY IIEQAGGLAT TGTQPVLDVK PEAIHQRVPL ILGSPEDVQE YLTCVQKNQA GS | |
Greater than 90.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The FBP2 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |