missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FARSB Protein
A cDNA sequence encoding the FARSB was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11852-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
FARSB Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMPTVSVK RDLLFQALGR TYTDEEFDEL CFEFGLELDE ITSEKEIISK EQGNVKAAGA SDVVLYKIDV PANRYDLLCL EGLVRGLQVF KERIKAPVYK RVMPDGKIQK LIITEETAKI RPFAVAAVLR NIKFTKDRYD SFIELQEKLH QNICRKRALV AIGTHDLDTL SGPFTYTAKR PSDIKFKPLN KTKEYTACEL MNIYKTDNHL KHYLHIIENK PLYPVIYDSN GVVLSMPPII NGDHSRITVN TRNIFIECTG TDFTKAKIVL DIIVTMFSEY CENQFTVEAA EVVFPNGKSH TFPELAYRKE MVRADLINKK VGIRETPENL AKLLTRMYLK SEVIGDGNQI EIEIPPTRAD IIHACDIVED AAIAYGYNNI QMTLPKTYTI ANQFPLNKLT ELLRHDMAAA GFTEALTFAL CSQEDIADKL GVDISATKAV HISNPKTAEF QVARTTLLPG LLKTIAANRK MPLPLKLFEI SDIVIKDSNT DVGAKNYRHL CAVYYNKNPG FEIIHGLLDR IMQLLDVPPG EDKGGYVIKA SEGPAFFPGR CAEIFARGQS VGKLGVLHPD VITKFELTMP CSSLEINVGP FL | |
Greater than 85.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
FARSB protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol. |