missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FARS2 Protein
A cDNA sequence encoding the FARS2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11851-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
FARS2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MPAAECATQR APGSVVELLG KSYPQDDHSN LTRKVLTRVG RNLHNQQHHP LWLIKERVKE HFYKQYVGRF GTPLFSVYDN LSPVVTTWQN FDSLLIPADH PSRKKGDNYY LNRTHMLRAH TSAHQWDLLH AGLDAFLVVG DVYRRDQIDS QHYPIFHQLE AVRLFSKHEL FAGIKDGESL QLFEQSSRSA HKQETHTMEA VKLVEFDLKQ TLTRLMAHLF GDELEIRWVD CYFPFTHPSF EMEINFHGEW LEVLGCGVME QQLVNSAGAQ DRIGWAFGLG LERLAMILYD IPDIRLFWCE DERFLKQFCV SNINQKVKFQ PLSKYPAVIN DISFWLPSEN YAENDFYDLV RTIGGDLVEK VDLIDKFVHP KTHKTSHCYR ITYRHMERTL SQREVRHIHQ ALQEAAVQLL GVEGRF | |
Greater than 90.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
FARS2 solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 30% glycerol, 1mM EDTA and 0.1M NaCl. |