missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human FAM150A Protein

A DNA sequence encoding the Homo sapiens (Human) FAM150A, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP7769-ec-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16050681

  • 2645.00€ / 1 miligramo


Fecha estimada de envío
19-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_997296.1
FAM150A Protein
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
389658
Greater than 90% as determined by SDS-PAGE.
Research Use Only
FAM150A
Recombinant Protein
E. coli
RPRGRRGARV TDKEPKPLLF LPAAGAGRTP SGSRSAEIFP RDSNLKDKFI KHFTGPVTFS PECSKHFHRL YYNTRECSTP AYYKRCARLL TRLAVSPLCS QT
Videos
Documentos