missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human FAM107B Protein

A cDNA sequence encoding the FAM107B was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP11842-5ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15928558

  • 107.00€ / 5 microgramos


Fecha estimada de envío
02-07-2020
Añadir a la cesta

Especificaciones

Especificaciones

FAM107B Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The FAM107B solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT.
Greater than 95% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMAEPDYI EDDNPELIRP QKLINPVKTS RNHQDLHREL LMNQKRGLAP QNKPELQKVM EKRKRDQVIK QKEEEAQKKK SDLEIELLKR QQKLEQLELE KQKLQEEQEN APEFVKVKGN LRRTGQEVAQ AQES
Videos
Documentos