missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FADD Protein
A cDNA sequence encoding the FADD was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11839-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
FADD Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHHGMASMTGGQQ MGRDLYDDDD KDRWGSMDPF LVLLHSVSSS LSSSELTELK FLCLGRVGKR KLERVQSGLD LFSMLLEQND LEPGHTELLR ELLASLRRHD LLRRVDDFEA GAAAGAAPGE EDLCAAFNVI CDNVGKDWRR LARQLKVSDT KIDSIEDRYP RNLTERVRES LRIWKNTEKE NATVAHLVGA LRSCQMNLVA DLVQEVQQAR DLQNRSGAMS PMSWNSDAST SEAS | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The FADD protein solution contains 20mM Tris-HCl, pH 8.0, and 10% glycerol. |