missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human EZR / VIL2 / Ezrin Protein

A DNA sequence encoding the Homo sapiens (Human) EZR / VIL2 / Ezrin, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6006-ye-100ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15919309

  • 530.00€ / 100 microgramos


Fecha estimada de envío
16-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_001104547.1
EZR / VIL2 / Ezrin Protein
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
7430
Greater than 90% as determined by SDS-PAGE.
Research Use Only
EZR
Recombinant Protein
Yeast
MPKPINVRVT TMDAELEFAI QPNTTGKQLF DQVVKTIGLR EVWYFGLHYV DNKGFPTWLK LDKKVSAQEV RKENPLQFKF RAKFYPEDVA EELIQDITQK LFFLQVKEGI LSDEIYCPPE TAVLLGSYAV QAKFGDYNKE VHKSGYLSSE RLIPQRVMDQ HKLTRDQWED RIQVWHAEHR GMLKDNAMLE YLKIAQDLEM YGINYFEIKN KKGTDLWLGV DALGLNIYEK DDKLTPKIGF PWSEIRNISF N
Videos