missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Eyes absent homolog 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Eyes absent homolog 2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6005-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_005235.3 | |
Eyes absent homolog 2 Protein | |
Research Use Only | |
EYA2 | |
Recombinant Protein | |
E. coli | |
MVELVISPSL TVNSDCLDKL KFNRADAAVW TLSDRQGITK SAPLRVSQLF SRSCPRVLPR QPSTAMAAYG QTQYSAGIQQ ATPYTAYPPP AQAYGIPSFS TSPTGQSPYT YQMHGTTGFY QGGNGLGNAA GFGSVHQDYP SYPGFPQSQY PQYYGSSYNP PYVPASSICP SPLSTSTYVL QEASHNVPNQ SSESLAGEYN THNGPSTPAK EGDTDRPHRA SDGKLRGRSK RSSDPSPAGD NEIERVFVWD LDETIIIFHS LLTGTFASRY GKDTTTSVRI GLMMEEMIFN LADTHLFFND LEDCDQIHVD DVSSDDNGQD LSTYNFSADG FHSSAPGANL CLGSGVHGGV DWMRKLAFRY RRVKEMYNTY KNNVGGLIGT PKRETWLQLR AELEALTDLW LTHSLKALNL INSRPNCVNV LVTTTQLIPA LAKVLLYGLG SVFPIENIYS ATKTGKESCF ERIMQRFGRK AVYVVIGDGV EEEQGAKKHN MPFWRISCHA DLEALRHALE LEYL | |
Greater than 90% as determined by SDS-PAGE. |
2139 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |