missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EXOSC10 Protein
A cDNA sequence encoding the EXOSC10 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11818-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
EXOSC10 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
MAPPSTREPR VLSATSATKS DGEMVLPGFP DADSFVKFAL GSVVAVTKAS GGLPQFGDEY DFYRSFPGFQ AFCETQGDRL LQCMSRVMQY HGCRSNIKDR SKVTELEDKF DLLVDANDVI LERVGILLDE ASGVNKNQQP VLPAGLQVPK TVVSSWNRKA AEYGKKAKSE TFRLLHAKNI IRPQLKFREK IDNSNTPFLP KIFIKPNAQK PLPQALSKER RERPQDRPED LDVPPALADF IHQQRTQQVE QDMFAHPYQY ELNHFTPADA VLQKPQPQLY RPIEETPCHF ISSLDELVEL NEKLLNCQEF AVDLEHHSYR SFLGLTCLMQ ISTRTEDFII DTLELRSDMY ILNESLTDPA IVKVFHGADS DIEWLQKDFG LYVVNMFDTH QAARLLNLGR HSLDHLLKLY CNVDSNKQYQ LADWRIRPLP EEMLSYARDD THYLLYIYDK MRLEMWERGN GQPVQLQVVW QRSRDICLKK FIKPIFTDES YLELYRKQKK HLNTQQLTAF QLLFAWRDKT ARREDESYGY VLPNHMMLKI AEELPKEPQG IIACCNPVPP LVRQQINEMH LLIQQAREMP LLKSEVAAGV KKSGPLPSAE RLENVLFGPH DCSHAPPDGY PIIPTSGSVP VQKQASLFPD EKEDNLLGTT CLIATAVITL FNEPSAEDSK KGPLTVAQKK AQNIMESFEN PFRMFLPSLG HRAPVSQAAK FDPSTKIYEI SNRWKLAQVQ VQKDSKEAVK KKAAEQTAAR EQAKEACKAA AEQAISVRQQ VVLENAAKKR ERATSDPRTT EQKQEKKRLK ISKKPKDPEP PEKEFTPYDY SQSDFKAFAG NSKSKVSSQF DPNKQTPSGK KCIAAKKIKQ SVGNKSMSFP TGKSDRGFRY NWPQR | |
Greater than 70% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
EXOSC10 is supplied at a concentration of 0.8 mg/ml in 20mM HEPES, pH 7.5, 100mM NaCl, 0.01mM EDTA, 0.02% SDS. |