missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human EXOSC1 Protein

A cDNA sequence encoding the EXOSC1 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP11817-2ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15908478

  • 103.00€ / 2 microgramos


Fecha estimada de envío
20-08-2019
Añadir a la cesta

Especificaciones

Especificaciones

51013
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
EXOSC1
Human
His
EXOSC1 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT.
EXOSC1 Protein
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMAPPVRY CIPGERLCNL EEGSPGSGTY TRHGYIFSSL AGCLMKSSEN GALPVVSVVR ETESQLLPDV GAIVTCKVSS INSRFAKVHI LYVGSMPLKN SFRGTIRKED VRATEKDKVE IYKSFRPGDI VLAKVISLGD AQSNYLLTTA ENELGVVVAH SESGIQMVPI SWCEMQCPKT HTKEFRKVAR VQPEFLQT
Videos
Documentos