missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Estrogen Receptor 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Estrogen Receptor 1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5998-ye-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15909249

  • 253.00€ / 50 microgramos


Fecha de envío estimada
del proveedor 22-05-2019
Añadir a la cesta

Especificación

Especificación

NP_000116.2
Estrogen Receptor 1 Protein
50 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
2099
Greater than 80% as determined by SDS-PAGE.
Research Use Only
ESR1
Recombinant Protein
Yeast
SGMALLHQIQ GNELEPLNRP QLKIPLERPL GEVYLDSSKP AVYNYPEGAA YEFNAAAAAN AQVYGQTGLP YGPGSEAAAF GSNGLGGFPP LNSVSPSPLM LLHPPPQLSP FLQPHGQQVP YYLENEPSGY TVREAGPPAF YRPNSDNRRQ GGRERLASTN DKGSMAMESA KETRYCAVCN DYASGYHYGV WSCEGCKAFF KRSIQGHNDY MCPATNQCTI DKNRRKSCQA CRLRKCYEVG MMKGGIRKDR RGGRMLKHKR QRDDGEGRGE VGSAGDMRAA NLWPSPLMIK RSKKNSLALS LTADQMVSAL LDAEPPILYS EYDPTRPFSE ASMMGLLTNL ADRELVHMIN WAKRVPGFVD LTLHDQVHLL ECAWLEILMI GLVWRSMEHP GKLLFAPNLL LDRNQGKCVE GMVEIFDMLL ATSSRFRMMN LQGEEFVCLK SIILLNSGVY TFLSSTLKSL EEKDHIHRVL DKITDTLIHL MAKAGLTLQQ QHQRLAQLLL ILSHIRHMSN KGMEHLYSMK CKNVVPLYDL LLEMLDAHRL HAPTSRGGAS VEETDQSHLA TAGSTSSHSL QKYYITGEAE GFPATV
Videos