missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Esterase D Protein
A cDNA sequence encoding the Esterase D was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11811-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
2098 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MALKQISSNK CFGGLQKVFE HDSVELNCKM KFAVYLPPKA ETGKCPALYW LSGLTCTEQN FISKSGYHQS ASEHGLVVIA PDTSPRGCNI KGEDESWDFG TGAGFYVDAT EDPWKTNYRM YSYVTEELPQ LINANFPVDP QRMSIFGHSM GGHGALICAL KNPGKYKSVS AFAPICNPVL CPWGKKAFSG YLGTDQSKWK AYDATHLVKS YPGSQLDILI DQGKDDQFLL DGQLLPDNFI AACTEKKIPV VFRLQEDYDH SYYFIATFIT DHIRHHAKYL NA | |
Greater than 95.0% as determined by SDS-PAGE. |
Esterase D Protein | |
Research Use Only | |
ESD | |
Human | |
His | |
The Esterase-D solution contains 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol. |