Learn More
enQuireBio™ Recombinant Human Esterase D Protein
A cDNA sequence encoding the Esterase D was constructed and used to recombinantly synthesize the protein.
137.00€ - 2725.00€
Specifications
ID de gen (Entrez) 2098
Nombre Esterase D Protein
Estado normativo Research Use Only
Concentración de endotoxinas Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Símbolo de gen ESD
View More Specs
Products 3
Product Code Brand Cantidad Price Quantity & Availability  
Product Code Brand Cantidad Price Quantity & Availability  
15908458
View Documents Promotion Details
enQuireBio™
QP11811-10UG
10 μg
137.00€
10 microgramos
View alternative products
Save Up to 
»
Fecha estimada de envío: 28-05-2024
para ver el stock
15928458
View Documents Promotion Details
enQuireBio™
QP11811-50UG
50 μg
212.00€
50 microgramos
View alternative products
Save Up to 
»
Fecha estimada de envío: 28-05-2024
para ver el stock
15918458
View Documents Promotion Details
enQuireBio™
QP11811-1MG
1 mg
2725.00€
1 miligramo
View alternative products
Save Up to 
»
Fecha estimada de envío: 28-05-2024
para ver el stock
Description
Description
Specifications
Specifications
2098
Research Use Only
ESD
Human
His
The Esterase-D solution contains 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol.
Esterase D Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MALKQISSNK CFGGLQKVFE HDSVELNCKM KFAVYLPPKA ETGKCPALYW LSGLTCTEQN FISKSGYHQS ASEHGLVVIA PDTSPRGCNI KGEDESWDFG TGAGFYVDAT EDPWKTNYRM YSYVTEELPQ LINANFPVDP QRMSIFGHSM GGHGALICAL KNPGKYKSVS AFAPICNPVL CPWGKKAFSG YLGTDQSKWK AYDATHLVKS YPGSQLDILI DQGKDDQFLL DGQLLPDNFI AACTEKKIPV VFRLQEDYDH SYYFIATFIT DHIRHHAKYL NA
Greater than 95.0% as determined by SDS-PAGE.
Videos
SDS
Documents
Documents
Product Certifications