missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ERP27 Protein
A cDNA sequence encoding the ERP27 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11808-2ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
ERP27 Protein | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The ERP27 solution (1 mg/ml) contains 20mM Tris buffer (pH 8.0), 10% glycerol and 1mM DTT. |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MEVEKSSDGP GAAQEPTWLT DVPAAMEFIA ATEVAVIGFF QDLEIPAVPI LHSMVQKFPG VSFGISTDSE VLTHYNITGN TICLFRLVDN EQLNLEDEDI ESIDATKLSR FIEINSLHMV TEYNPVTVIG LFNSVIQIHL LLIMNKASPE YEENMHRYQK AAKLFQGKIL FILVDSGMKE NGKVISFFKL KESQLPALAI YQTLDDEWDT LPTAEVSVEH VQNFCDGFLS GKLLKENRES EGKTPKVEL |