Learn More
enQuireBio™ Recombinant Human Eptifibatide Protein
Recombinant Protein
Marca:  enQuireBio™ QP11802-5mg
Detalles adicionales : Peso : 0.01000kg
 Ver más versiones de este producto
Código de producto. 15918428
  • 103.00€ / 5 miligramos
Fecha estimada de envío
22-07-2019
Añadir a la cesta
Especificaciones
Especificaciones
Eptifibatide Protein
5 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
Greater than 98.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
Eptifibatide was lyophilized with no additives.
Videos