missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Epoxide hydrolase 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Epoxide hydrolase 1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5985-ec-100ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15929149

  • 406.00€ / 100 microgramos


Fecha estimada de envío
23-03-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_000111.1
Epoxide hydrolase 1 Protein
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
2052
Greater than 80% as determined by SDS-PAGE.
Research Use Only
EPHX1
Recombinant Protein
E. coli
MWLEILLTSV LGFAIYWFIS RDKEETLPLE DGWWGPGTRS AAREDDSIRP FKVETSDEEI HDLHQRIDKF RFTPPLEDSC FHYGFNSNYL KKVISYWRNE FDWKKQVEIL NRYPHFKTKI EGLDIHFIHV KPPQLPAGHT PKPLLMVHGW PGSFYEFYKI IPLLTDPKNH GLSDEHVFEV ICPSIPGYGF SEASSKKGFN SVATARIFYK LMLRLGFQEF YIQGGDWGSL ICTNMAQLVP SHVKGLHLNM ALVLSNFSTL TLLLGQRFGR FLGLTERDVE LLYPVKEKVF YSLMRESGYM HIQCTKPDTV GSALNDSPVG LAAYILEKFS TWTNTEFRYL EDGGLERKFS LDDLLTNVML YWTTGTIISS QRFYKENLGQ GWMTQKHERM KVYVPTGFSA FPFELLHTPE KWVRFKYPKL ISYSYMVRGG HFAAFEEPEL LAQDIRKFLS VLERQ
Videos
Documentos