missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Epiregulin / EREG Protein

A DNA sequence encoding the Human Epiregulin / EREG, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10397-ec-25ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15923728

  • 299.00€ / 25 microgramos


Fecha de envío estimada
del proveedor 20-05-2019
Añadir a la cesta

Especificación

Especificación

NP_001423.1
Epiregulin / EREG Protein
25 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
VAQVSITKCS SDMNGYCLHG QCIYLVDMSQ NYCRCEVGYT GVRCEHFFL
2069
Greater than 97% as determined by SDS-PAGE and HPLC.
Research Use Only
EREG
The ED50 as determined by a cell proliferation assay using murine Balb/c 3T3 cells is less than 2 ng/ml, corresponding to a specific activity of > 5.0 M 105 IU/mg.
Human
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos