missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Enterokinase Protein

A cDNA sequence encoding the Enterokinase was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP11792-500IU

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15918388

  • 1337.00€ / 500 unidades


Fecha estimada de envío
15-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

Enterokinase Protein
500 IU
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
50mM Tris-HCl, pH 8.0, 0.5M NaCl, 1mM CaCl2 and 50% glycerol.
Greater than 95.0% as determined by HPLC-SEC.
Research Use Only
Recombinant Protein
E. coli
MGSKRGISSR HHSLSSYEIM FAALFAILVV LCAGLIAVSC LTIKESQRGA ALGQSHEARA TFKITSGVTY NPNLQDKLSV DFKVLAFDLQ QMIDEIFLSS NLKNEYKNSR VLQFENGSII VVFDLFFAQW VSDENVKEEL IQGLEANKSS QLVTFHIDLN SVDILDKLTT TSHLATPGNV SIECLPGSSP CTDALTCIKA DLFCDGEVNC PDGSDEDNKM CATVCDGRFL LTGSSGSFQA THYPKPSETS VVCQWIIRVN QGLSIKLSFD DFNTYYTDIL DIYEGVGSSK ILRASIWETN PGTIRIFSNQ VTATFLIESD ESDYVGFNAT YTAFNSSELN NYEKINCNFE DGFCFWVQDL NDDNEWERIQ GSTFSPFTGP NFDHTFGNAS GFYISTPTGP GGRQERVGLL SLPLDPTLEP ACLSFWYHMY GENVHKLSIN ISNDQNMEKT VFQKEGNYGD NWNYGQVTLN ETVKFKVAFN AFKNKILSDI ALDDISLTYG ICNGSLYPEP TLVPTPPPEL PTDCGGPFEL WEPNTTFSST NFPNSYPNLA FCVWILNAQK GKNIQLHFQE FDLENINDVV EIRDGEEADS LLLAVYTGPG PVKDVFSTTN RMTVLLITND VLARGGFKAN FTTGYHLGIP EPCKADHFQC KNGECVPLVN LCDGHLHCED GSDEADCVRF FNGTTNNNGL VRFRIQSIWH TACAENWTTQ ISNDVCQLLG LGSGNSSKPI FPTDGGPFVK LNTAPDGHLI LTPSQQCLQD SLIRLQCNHK SCGKKLAAQD ITPKIVGGSN AKEGAWPWVV GLYYGGRLLC GASLVSSDWL VSAAHCVYGR NLEPSKWTAI LGLHMKSNLT SPQTVPRLID EIVINPHYNR RRKDNDIAMM HLEFKVNYTD YIQPICLPEE NQVFPPGRNC SIAGWGTVVY QGTTANILQE ADVPLLSNER CQQQMPEYNI TENMICAGYE EGGIDSCQGD SGGPLMCQEN NRWFLAGVTS FGYKCALPNR PGVYARVSRF TEWIQSFLH
Videos