missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ENPEP / Aminopeptidase A Protein
A DNA sequence encoding the Homo sapiens (Human) ENPEP / Aminopeptidase A, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8812-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001968.3 | |
ENPEP / Aminopeptidase A Protein | |
Research Use Only | |
ENPEP | |
Recombinant Protein | |
E. coli | |
YFQGQVKPIA DSLGWNDAGD HVTKLLRSSV LGFACKMGDR EALNNASSLF EQWLNGTVSL PVNLRLLVYR YGMQNSGNEI SWNYTLEQYQ KTSLAQEKEK LLYGLASVKN VTLLSRYLDL LKDTNLIKTQ DVFTVIRYIS YNSYGKNMAW NWIQLNWDYL VNRYTLNNRN LGRIVTIAEP FNTELQLWQM ESFFAKYPQA GAGEKPREQV LETVKNNIEW LKQHRNTIRE W | |
Greater than 90% as determined by SDS-PAGE. |
2028 | |
50 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |