missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ENO3 / beta-enolase Protein

A DNA sequence encoding the Homo sapiens (Human) ENO3 / beta-enolase, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP1054-ec-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15934188

  • 234.00€ / 50 microgramos


Fecha estimada de envío
24-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_001180432.1
ENO3 / beta-enolase Protein
50 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
2027
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ENO3
Recombinant Protein
E. coli
FAREILDSRG NPTVEVDLHT AKGRFRAAVP SGASTGIYEA LELRDGDKGR YLGKGVLKAV ENINNTLGPA LLQKKLSVVD QEKVDKFMIE LDGTENKSKF GANAILGVSL AVCKAGAAEK GVPLYRHIAD LAGNPDLILP VPAFNVINGG SHAGNKLAMQ EFMILPVGAS SFKEAMRIGA EVYHHLKGVI KAKYGKDATN VGDEGGFAPN ILENNEALEL LKTAIQAAGY PDKVVIGMDV AASEFYRNGK YDLDFKSPDD PARHITGEKL GELYKSFIKN YPVVSIEDPF DQDDWATWTS FLSGVNIQIV GDDLTVTNPK RIAQAVEKKA CNCLLLKVNQ IGSVTESIQA CKLAQSNGWG VMVSHRSGET EDTFIADLVV GLCTGQIKTG APCRSERLAK YNQLMRIEEA LGDKAIFAGR KFRNPK
Videos
Documentos