missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Endophilin-B2 Protein
A DNA sequence encoding the Homo sapiens (Human) Endophilin-B2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8092-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001273974.1 | |
Endophilin-B2 Protein | |
Research Use Only | |
SH3GLB2 | |
Recombinant Protein | |
E. coli | |
MDFNMKKLAS DAGIFFTRAV QFTEEKFGQA EKTELDAHFE NLLARADSTK NWTEKILRQT EVLLQPNPSA RVEEFLYEKL DRKVPSRVTN GELLAQYMAD AASELGPTTP YGKTLIKVAE AEKQLGAAER DFIHTASISF LTPLRNFLEG DWKTISKERR LLQNRRLDLD ACKARLKKAK AAEAKATTVP DFQETRPRNY ILSASASALW NDEVDKAEQE LRVAQTEFDR QAEVTRLLLE GISSTHVNHL RCLHEFVKSQ TTYYAQCYRH MLDLQKQLGR FPGTFVGTTE PASPPLSSTS PTTAAATMPV VPSVASLAPP GEASLCLEEV APPASGTRKA RVLYDYEAAD SSELALLADE LITVYSLPGM DPDWLIGERG NKKGKVPVTY LELLS | |
Greater than 90% as determined by SDS-PAGE. |
56904 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |