missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Endophilin-B2 Protein

A DNA sequence encoding the Homo sapiens (Human) Endophilin-B2, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8092-ec-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16052621

  • 160.00€ / 10 microgramos


Fecha estimada de envío
25-06-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_001273974.1
Endophilin-B2 Protein
10 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
56904
Greater than 90% as determined by SDS-PAGE.
Research Use Only
SH3GLB2
Recombinant Protein
E. coli
MDFNMKKLAS DAGIFFTRAV QFTEEKFGQA EKTELDAHFE NLLARADSTK NWTEKILRQT EVLLQPNPSA RVEEFLYEKL DRKVPSRVTN GELLAQYMAD AASELGPTTP YGKTLIKVAE AEKQLGAAER DFIHTASISF LTPLRNFLEG DWKTISKERR LLQNRRLDLD ACKARLKKAK AAEAKATTVP DFQETRPRNY ILSASASALW NDEVDKAEQE LRVAQTEFDR QAEVTRLLLE GISSTHVNHL RCLHEFVKSQ TTYYAQCYRH MLDLQKQLGR FPGTFVGTTE PASPPLSSTS PTTAAATMPV VPSVASLAPP GEASLCLEEV APPASGTRKA RVLYDYEAAD SSELALLADE LITVYSLPGM DPDWLIGERG NKKGKVPVTY LELLS
Videos
Documentos