missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human En Protein
A cDNA sequence encoding the En was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11784-2ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
2022 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MDRGTLPLAV ALLLASCSLS PTSLAETVHC DLQPVGPERG EVTYTTSQVS KGCVAQAPNA ILEVHVLFLE FPTGPSQLEL TLQASKQNGT WPREVLLVLS VNSSVFLHLQ ALGIPLHLAY NSSLVTFQEP PGVNTTELPS FPKTQILEWA AERGPITSAA ELNDPQSILL RLGQAQGSLS FCMLEASQDM GRTLEWRPRT PALVRGCHLE GVAGHKEAHI LRVLPGHSAG PRTVTVKVEL SCAPGDLDAV LILQGPPYVS WLIDANHNMQ IWTTGEYSFK IFPEKNIRGF KLPDTPQGLL GEARMLNASI VASFVELPLA SIVSLHASSC GGRLQTSPAP IQTTPPKDTC SPELLMSLIQ TKCADDAMTL VLKKELVAHL KCTITGLTFW DPSCEAEDRG DKFVLRSAYS SCGMQVSASM ISNEAVVNIL SSSSPQRKKV HCLNMDSLSF QLGLYLSPHF LQASNTIEPG QQSFVQVRVS PSVSEFLLQL DSCHLDLGPE GGTVELIQGR AAKGNCVSLL SPSPEGDPRF SFLLHFYTVP IPKTGTLSCT VALRPKTGSQ DQEVHRTVFM RLNIISPDLS GCTSKGLVLP AVLGITFGAF LIGALLTAAL WYIYSHTRSP SKREPVVAVA APASSESSST NHSIGSTQST PCSTSSMA | |
Greater than 90.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
En Protein | |
Research Use Only | |
ENG | |
Human | |
Untagged | |
Endoglin solution in 50mM Tris-Acetate, pH 7.5, 1mM EDTA and 20% Glycerol. |