missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EML2 Protein
A DNA sequence encoding the Homo sapiens (Human) EML2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5982-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001180197.1 | |
EML2 Protein | |
Research Use Only | |
EML2 | |
Recombinant Protein | |
E. coli | |
MSSFGAGKTK EVIFSVEDGS VKMFLRGRPV PMMIPDELAP TYSLDTRSEL PSCRLKLEWV YGYRGRDCRA NLYLLPTGEI VYFVASVAVL YSVEEQRQRH YLGHNDDIKC LAIHPDMVTI ATGQVAGTTK EGKPLPPHVR IWDSVSLSTL HVLGLGVFDR AVCCVGFSKS NGGNLLCAVD ESNDHMLSVW DWAKETKVVD VKCSNEAVLV ATFHPTDPTV LITCGKSHIY FWTLEGGSLS KRQGLFEKHE KPKYVLCVTF LEGGDVVTGD SGGNLYVWGK GGNRITQAVL GAHDGGVFGL CALRDGTLVS GGGRDRRVVL WGSDYSKLQE VEVPEDFGPV RTVAEGHGDT LYVGTTRNSI LQGSVHTGFS LLVQGHVEEL WGLATHPSRA QFVTCGQDKL VHLWSSDSHQ PLWSRIIE | |
Greater than 90% as determined by SDS-PAGE. |
24139 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |