missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Elongation factor Tu, mitochondrial Protein

A DNA sequence encoding the Homo sapiens (Human) Elongation factor Tu, mitochondrial, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8402-ec-200ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16054411

  • 675.00€ / 200 microgramos


Fecha estimada de envío
26-06-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_003312.3
Elongation factor Tu, mitochondrial Protein
200 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
7284
Greater than 90% as determined by SDS-PAGE.
Research Use Only
TUFM
Recombinant Protein
E. coli
EAKKTYVRDK PHVNVGTIGH VDHGKTTLTA AITKILAEGG GAKFKKYEEI DNAPEERARG ITINAAHVEY STAARHYAHT DCPGHADYVK NMITGTAPLD GCILVVAAND GPMPQTREHL LLARQIGVEH VVVYVNKADA VQDSEMVELV ELEIRELLTE FGYKGEETPV IVGSALCALE GRDPELGLKS VQKLLDAVDT YIPVPARDLE KPFLLPVEAV YSVPGRGTVV TGTLERGILK KGDEC
Videos
Documentos